Få mere at vide
2-[[3,4,6-Tri-O-acetyl-2-(acetylamino)-2-deoxy-Beta-D-galactopyranosyl]oxy]acetic Acid, TRC
Artikelnummer. 30453476
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30453476 250 mg
30452267 500 mg
30451340 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30453476 Leverandør Toronto Research Chemicals Leverandørnr. TRCT992240250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1159409-18-3
Molekylær formel C16H23NO11
Molekylvægt (g/mol) 405.35
Synonym 2-(((2R,3R,4R,5R,6R)-3-Acetamido-4,5-diacetoxy-6-(acetoxymethyl)tetrahydro-2H-pyran-2-yl)oxy)acetic Acid
IUPAC navn 2-[(2R,3R,4R,5R,6R)-3-acetamido-4,5-diacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxyacetic acid
SMIL CC(=O)N[C@H]1[C@H](OCC(=O)O)O[C@H](COC(=O)C)[C@H](OC(=O)C)[C@@H]1OC(=O)C
Tekniske data
Kemisk navn eller materiale 2-((3,4,6-Tri-O-acetyl-2-(acetylamino)-2-deoxy-beta-D-galactopyranosyl)oxy)acetic Acid
Mængde 250 mg
InChI formel InChI=1S/C16H23NO11/c1-7(18)17-13-15(27-10(4)21)14(26-9(3)20)11(5-24-8(2)19)28-16(13)25-6-12(22)23/h11,13-16H,5-6H2,1-4H3,(H,17,18)(H,22,23)/t11-,13-,14+,15-,16-/m1/s1
Formel vægt 405.1271