Få mere at vide
2,3,4,6-Tetra-O-benzyl-Alpha-D-glucopyranosyl Bromide 90%Stabilized with Calcium Carbonate, TRC
Artikelnummer. 30415779
Skift visning
Mængde:
500 mg
2.5 g
5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30415779 500 mg -
30416427 5 g -
30419209 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30415779 Leverandør Toronto Research Chemicals Leverandørnr. TRCT289205500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4196-35-4
Molekylær formel C34H35BrO5
Molekylvægt (g/mol) 603.54
Synonym 2,3,4,6-Tetrakis-O-(phenylmethyl)-alpha-D-glucopyranosyl Bromide,, 2,3,4,6-Tetra-O-benzyl-glucopyranosyl Bromide, alpha-D-2,3,4,6-Tetra-O-benzyl-glucopyranosyl Bromide, 2,3,4,6-Tetra-O-benzyl-1-bromo-1-deoxy-alpha-D-glucopyranose, 2,3,4,6-Tetra-O-benzyl-alpha-D-bromoglucopyranoside, 2,3,4,6-Tetra-O-benzyl-alpha-D-glucopyranosyl Bromide, Tetra-O-benzyl-alpha-D-glucopyranosyl Bromide
IUPAC navn (2R,3R,4S,5R,6R)-2-bromo-3,4,5-tris(phenylmethoxy)-6-(3-phenylpropyl)oxane
SMIL Br[C@H]1O[C@H](CCCc2ccccc2)[C@@H](OCc3ccccc3)[C@H](OCc4ccccc4)[C@H]1OCc5ccccc5
Tekniske data
Kemisk navn eller materiale 2,3,4,6-Tetra-O-benzyl-alpha-D-glucopyranosyl Bromide
Mængde 500 mg
InChI formel InChI=1S/C35H37BrO4/c36-35-34(39-26-30-20-11-4-12-21-30)33(38-25-29-18-9-3-10-19-29)32(37-24-28-16-7-2-8-17-28)31(40-35)23-13-22-27-14-5-1-6-15-27/h1-12,14-21,31-35H,13,22-26H2/t31-,32-,33+,34-,35+/m1/s1
Formel vægt 600.1875