Få mere at vide
2,2'-Dinitrodibenzyl, TRC
Artikelnummer. 30134406
Skift visning
Mængde:
500 mg
1 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134406 1 g
30134315 25 g
30010818 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134406 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4798151G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 16968-19-7
Molekylær formel C14H12N2O4
Molekylvægt (g/mol) 272.26
Synonym 1,1'-(1,2-Ethanediyl)bis[2-nitrobenzene], 2,2'-Dinitrobibenzyl, 1,2-Bis(o-nitrophenyl)ethane, 1,2-Di(o-nitrophenyl)ethane, Bis(o-nitrobenzyl), NSC 85868, o,o'-Dinitrodibenzyl
SMIL [O-][N+](=O)c1ccccc1CCc2ccccc2[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale 2,2'-Dinitrodibenzyl
Mængde 1 g
InChI formel InChI=1S/C14H12N2O4/c17-15(18)13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)16(19)20/h1-8H,9-10H2
Formel vægt 272.0797