Få mere at vide
2,2-Bis(4-hydroxyphenyl)propanol, TRC
Artikelnummer. 30009944
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009944 25 mg
30012434 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30009944 Leverandør Toronto Research Chemicals Leverandørnr. TRCB44736025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 142648-65-5
Molekylær formel C15 H16 O3
Molekylvægt (g/mol) 244.28
Synonym 4-Hydroxy-β-(4-hydroxyphenyl)-β-methyl-benzeneethanol
IUPAC navn 4-[2-hydroxy-1-(4-hydroxyphenyl)-1-methyl-ethyl]phenol
SMIL CC(CO)(c1ccc(O)cc1)c2ccc(O)cc2
Tekniske data
Kemisk navn eller materiale 2,2-Bis(4-hydroxyphenyl)propanol
Mængde 25 mg
InChI formel InChI=1S/C15H16O3/c1-15(10-16,11-2-6-13(17)7-3-11)12-4-8-14(18)9-5-12/h2-9,16-18H,10H2,1H3
Formel vægt 244.1099