Få mere at vide
2,2,4-Trimethylpentane, ≥99.5% (GC), Honeywell Riedel-de Haën™
Artikelnummer. 15625880
Skift visning
Mængde:
1 L
2.5 L
Emballage:
Glass Bottle
Pakningsstørrelse:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Emballage unitSize
15625880 1 L Glass Bottle 1 L
15635880 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Artikelnummer. 15625880 Leverandør Honeywell-Riedel-de Haen Leverandørnr. 590451L
Request Bulk or Custom Format
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Puriss. p.a., ACS Reagent,≥99.5% (GC)
ACS GENERAL USE SOLVENTS
  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 540-84-1
Molekylær formel C8H18
Molekylvægt (g/mol) 114.232
MDL nummer MFCD00008943
InChI nøgle NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC navn 2,2,4-trimethylpentane
SMIL CC(C)CC(C)(C)C
Tekniske data
Kemisk navn eller materiale 2,2,4-Trimethylpentane
Smeltepunkt -107°C
Farve Colorless
Tæthed 0.692g/mL at 25°C
Mængde 1 L
Eksplosionsgrænse 0.06
Tændingspunkt 745°C
Emballage Glass Bottle
Brydningsindeks 1.39 to 1.392
Lineær formel (CH3)2CHCH2C(CH3)3
Damptryk 41mmHg (21°C)
FN nummer UN1262
Beilstein 1696876
Kogepunkt 98°C to 100°C
Formel vægt 114.23g/mol
Procent renhed ≥99.5% (GC)