Få mere at vide
2,2',4,4',5-Pentabromodiphenyl Ether (>80%)(Contains ~10% 2,2',4,4',5,5'-Hexabromomdiphenyl Ether), TRC
Artikelnummer. 30135887
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135887 50 mg
30132703 5 mg
17963805 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135887 Leverandør Toronto Research Chemicals Leverandørnr. TRCP23776550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 60348-60-9
Molekylær formel C12 H5 Br5 O
Molekylvægt (g/mol) 564.69
Synonym Benzene, 1,2,4-tribromo-5-(2,4-dibromophenoxy)-, 1,2,4-Tribromo-5-(2,4-dibromophenoxy)benzene, 2,2',4,4',5-Pentabromodiphenyl ether, 2,2',4,4',5-Pentabromodiphenyl oxide, BDE 99, PBDE 99, Tardex 50, Tardex 50L
IUPAC navn 1,2,4-tribromo-5-(2,4-dibromophenoxy)benzene
SMIL Brc1ccc(Oc2cc(Br)c(Br)cc2Br)c(Br)c1
Tekniske data
Kemisk navn eller materiale PBDE 99 (2,2',4,4',5-Pentabromodiphenyl Ether)
Mængde 50 mg
InChI formel InChI=1S/C12H5Br5O/c13-6-1-2-11(9(16)3-6)18-12-5-8(15)7(14)4-10(12)17/h1-5H
Formel vægt 559.6257