Learn More
(1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane, 97%
Description
It is used as an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used as an active pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Air sensitive. Store away from air, carbon dioxide, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep it refrigerated.

Specifications
Specifications
Melting Point | 74°C to 76°C |
Quantity | 5 g |
Beilstein | 6460311 |
Sensitivity | Air sensitive |
Solubility Information | Insoluble in water. |
Optical Rotation | −44° (c=1 in Chloroform) |
Formula Weight | 198.26 |
Percent Purity | 97% |
Chemical Name or Material | (1S,4S)-(-)-2-Boc-2,5-diazabicyclo[2.2.1]heptane |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
10136932 | Certificate of Analysis | 25/07/2023 | L20131.03,L20131.06,L20131.MD |
G18Q38 | Certificate of Analysis | 08/12/2021 | L20131.03,L20131.06 |
K23W010 | Certificate of Analysis | 08/12/2021 | L20131.03,L20131.06 |
H08R005 | Certificate of Analysis | 07/12/2021 | L20131.03,L20131.06 |
B23S027 | Certificate of Analysis | 07/12/2021 | L20131.03,L20131.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.