Få mere at vide
(1R,2S)-rel-1,2-Cyclohexanedimethanol, TRC
Artikelnummer. 30366445
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366445 10 g
30360478 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30366445 Leverandør Toronto Research Chemicals Leverandørnr. TRCC98705010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 15753-50-1
Molekylær formel C8H16O2
Molekylvægt (g/mol) 144.21
Synonym cis-1,2-cyclohexanedimethanol, (1R,2S)-rel-1,2-Cyclohexanedimethanol, (cis-2-Hydroxymethylcyclohexyl)methanol, 1,2-cis-Cyclohexanedimethanol, cis-1,2-Bis(hydroxymethyl)cyclohexane, cis-1,2-Cyclohexanedimethanol, cis-[2-(Hydroxymethyl)cyclohexane]methanol
IUPAC navn [(1R,2S)-2-(hydroxymethyl)cyclohexyl]methanol
SMIL OC[C@@H]1CCCC[C@@H]1CO
Tekniske data
Kemisk navn eller materiale (1R,2S)-rel-1,2-Cyclohexanedimethanol
Mængde 10 g
InChI formel InChI=1S/C8H16O2/c9-5-7-3-1-2-4-8(7)6-10/h7-10H,1-6H2/t7-,8+
Formel vægt 144.115