Få mere at vide
18α-Glycyrrhetinic acid, MedChemExpress
Artikelnummer. p-200098806
Encompass_Preferred
Skift visning
Mængde:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16449066 100 mg
16432376 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16449066 Leverandør Medchem Express Leverandørnr. HYN0375
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
MedChemExpress 18α-Glycyrrhetinic acid, a diet-derived compound, is an inhibitor of NF-kB and an activator of proteasome, which serves as pro-longevity and anti-aggregation factor in a multicellular organism. 18α-Glycyrrhetinic acid induces apoptosis.
18α-Glycyrrhetinic acid, a diet-derived compound, is an inhibitor of NF-kB and an activator of proteasome, which serves as pro-longevity and anti-aggregation factor in a multicellular organism. 18α-Glycyrrhetinic acid induces apoptosis.
Tekniske data
Kemisk navn eller materiale 18α-Glycyrrhetinic acid
CAS 1449-05-4
Farve White
Renhedsgrad noter Research
Molekylær formel C30H46O4
Mængde 100 mg
Opløselighedsinformation DMSO : 19.23 mg/mL (40.86 mM; Need ultrasonic)
SMIL CC1(C)[C@@H](O)CC[C@@]2(C)[C@@]1([H])CC[C@]([C@@](CC[C@]3(C)[C@]4([H])C[C@@](C)(C(O)=O)CC3)(C)C4=C5)(C)[C@]2([H])C5=O
Molekylvægt (g/mol) 470.68
Formel vægt 470.68
Procent renhed 95.4%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease