Få mere at vide
(16beta)-16-Methyl-17,21-bis(1-oxopropoxy)pregna-1,4,9(11)-triene-3,20-dione, TRC
Artikelnummer. 30004238
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30004238 50 mg
30008901 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30004238 Leverandør Toronto Research Chemicals Leverandørnr. TRCM29422550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 52092-12-3
Molekylær formel C28H36O6
Molekylvægt (g/mol) 468.58
Synonym Pregna-1,4,9(11)-triene-3,20-dione, 16-methyl-17,21-bis(1-oxopropoxy)-, (16β)-, (16β)-16-Methyl-17,21-bis(1-oxopropoxy)pregna-1,4,9(11)-triene-3,20-dione
SMIL CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)C3=CC[C@]12C
Tekniske data
Kemisk navn eller materiale (16-beta-)-16-Methyl-17,21-bis(1-oxopropoxy)pregna-1,4,9(11)-triene-3,20-dione
Mængde 50 mg
InChI formel InChI=1S/C28H36O6/c1-6-24(31)33-16-23(30)28(34-25(32)7-2)17(3)14-22-20-9-8-18-15-19(29)10-12-26(18,4)21(20)11-13-27(22,28)5/h10-12,15,17,20,22H,6-9,13-14,16H2,1-5H3/t17-,20+,22-,26-,27-,28-/m0/s1
Formel vægt 468.2512