Få mere at vide
(11Z)-11-Eicosenoic Acid, TRC
Artikelnummer. 30134124
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134124 10 mg
30132682 50 mg
30008772 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134124 Leverandør Toronto Research Chemicals Leverandørnr. TRCE47804510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5561-99-9
Molekylær formel C20 H38 O2
Molekylvægt (g/mol) 310.51
Synonym (Z)-11-Eicosenoic Acid, cis-11-Eicosenoic acid
IUPAC navn (Z)-icos-11-enoic acid
SMIL CCCCCCCC\C=C/CCCCCCCCCC(=O)O
Tekniske data
Kemisk navn eller materiale Gondoic Acid
Mængde 10 mg
InChI formel InChI=1S/C20H38O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h9-10H,2-8,11-19H2,1H3,(H,21,22)/b10-9-
Formel vægt 310.2872