Få mere at vide
11-Beta-Hydroxyandrosterone, TRC
Artikelnummer. 30007653
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007653 100 mg
30006346 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30007653 Leverandør Toronto Research Chemicals Leverandørnr. TRCH755000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57-61-4
Molekylær formel C19 H30 O3
Molekylvægt (g/mol) 306.44
Synonym Androstan-17-one, 3,11-dihydroxy-, (3α,5α,11β)-, 5α-Androstan-17-one, 3α,11β-dihydroxy- (8CI), (3α,5α,11β)-3,11-Dihydroxyandrostan-17-one, 11-Hydroxyandrosterone, 11-Oxyetiocholanolone, 11β-Hydroxyandrosterone, 3α,11β-Dihydroxy-5α-androstan-17-one, 3α,11β-Dihydroxy-5α-androstane-17-one, 5α-Androstane-3α,11β-diol-17-one, Ba 2685, NSC 56288
IUPAC navn (3R,5S,8S,9S,10S,11S,13S,14S)-3,11-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthren-17-one
SMIL C[C@]12CC[C@@H](O)C[C@@H]1CC[C@H]3[C@@H]4CCC(=O)[C@@]4(C)C[C@H](O)[C@H]23
Tekniske data
Kemisk navn eller materiale 11-beta-Hydroxyandrosterone
Mængde 100 mg
InChI formel InChI=1S/C19H30O3/c1-18-8-7-12(20)9-11(18)3-4-13-14-5-6-16(22)19(14,2)10-15(21)17(13)18/h11-15,17,20-21H,3-10H2,1-2H3/t11-,12+,13-,14-,15-,17+,18-,19-/m0/s1
Formel vægt 306.2195