Få mere at vide
1-Phenylsulfonyl-7-azaindole-3-boronic acid pinacol ester, 95%, Thermo Scientific™
Artikelnummer. 15437815
Skift visning
Mængde:
250 mg
Pakningsstørrelse:
250mg
Artikelnummer. Mængde unitSize
15437815 250 mg 250mg
1 options
Artikelnummer. 15437815 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H31505.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Phenylsulfonyl-7-azaindole-3-boronic acid pinacol ester
CAS 886547-94-0
Molekylær formel C19H21BN2O4S
MDL nummer MFCD09999194
Mængde 250 mg
Synonym 1-phenylsulfonyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrolo 2,3-b pyridine, 1-phenylsulfonyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 1-benzenesulfonyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrrolo 2,3-b pyridine, 1-benzenesulfonyl-1h-pyrrolo 2,3-b pyridine-3-boronic acid pinacol ester, 1-phenylsulfonyl-7-azaindole-3-boronic acid pinacol ester, 1-benzenesulfonyl-3-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 1-benzenesulfonyl-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, 1-benzenesulfonyl-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrrolo 2,3-b pyridine, ksc916a4p
InChI nøgle KZANVIJXSQABKR-UHFFFAOYSA-N
SMIL CC1(C)OB(OC1(C)C)C1=CN(C2=NC=CC=C12)S(=O)(=O)C1=CC=CC=C1
IUPAC navn 1-(benzenesulfonyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine
Molekylvægt (g/mol) 384.26
PubChem CID 45588137
Formel vægt 384.27
Procent renhed 95%