Få mere at vide
1-Phenylbiguanide hydrochloride, TRC
Artikelnummer. 30423871
Skift visning
Mængde:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30423871 250 mg
30426454 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30423871 Leverandør Toronto Research Chemicals Leverandørnr. TRCP319913250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 55-57-2
Molekylær formel C8H11N5 .HCl
Molekylvægt (g/mol) 213.67
Synonym Phenylbiguanide Monohydrochloride, N'-Phenylbiguanide Monohydrochloride, N-Phenylimidodicarbonimidic Diamide Monohydrochloride, N-Phenylimidodicarbonimidic Diamide Hydrochloride, 1-Phenylbiguanide Monohydrochloride
IUPAC navn 1-carbamimidoyl-3-phenylguanidine;hydrochloride
SMIL Cl.NC(=N)NC(=N)Nc1ccccc1
Tekniske data
Kemisk navn eller materiale 1-Phenylbiguanide hydrochloride
Mængde 250 mg
InChI formel InChI=1S/C8H11N5.ClH/c9-7(10)13-8(11)12-6-4-2-1-3-5-6;/h1-5H,(H6,9,10,11,12,13);1H
Formel vægt 213.0781