Learn More
1-n-Butyl-3-methylimidazolium bromide, 99% Product Code.: 11342179
1-n-Butyl-3-methylimidazolium bromide, 99%
Product Code. p-7029708
5 g, 5g
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
Product Code. 11342179
Brand: Thermo Scientific Alfa Aesar H27201.06
Request Bulk or Custom Format
CAS: 85100-77-2 | C8H15BrN2 | 219.126 g/mol
It is an ionic liquid used in a Heck reaction. It is also used in assessing the factors responsible for ionic liquid toxicity to aquatic organisms via quantitative structure-property relationship modeling
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an ionic liquid used in a Heck reaction. It is also used in assessing the factors responsible for ionic liquid toxicity to aquatic organisms via quantitative structure-property relationship modeling
Solubility
Miscible in water, methanol and dichloromethane. Immiscible in acetone, toluene, ethyl acetate and diethyl ether.
Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 85100-77-2
Molecular Formula C8H15BrN2
Molecular Weight (g/mol) 219.126
MDL Number MFCD03427611
InChI Key KYCQOKLOSUBEJK-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium bromide, 3-butyl-1-methyl-1h-imidazol-3-ium bromide, bmimbr, 1-n-butyl-3-methylimidazolium bromide, 1-butyl-3-methyl-1h-imidazol-3-ium bromide, bmim br, c4mim bromide, ksc491c7d, 1-butyl-3-methylimidazoliumbromideShow MoreShow Less
PubChem CID 2734236
IUPAC Name 1-butyl-3-methylimidazol-3-ium;bromide
SMILES CCCCN1C=C[N+](=C1)C.[Br-]
Specifications
Melting Point 65°C to 75°C
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Miscible in water,methanol and dichloromethane. Immiscible in acetone,toluene,ethyl acetate and diethyl ether.
Formula Weight 219.12
Percent Purity 99%
Chemical Name or Material 1-n-Butyl-3-methylimidazolium bromide
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number10254544 Certificate TypeCertificate of Analysis Date17/05/2025 Product CodeH27201.06,H27201.18
Lot Number10247680 Certificate TypeCertificate of Analysis Date25/08/2023 Product CodeH27201.06,H27201.18
Lot Number10241513 Certificate TypeCertificate of Analysis Date05/07/2022 Product CodeH27201.06,H27201.18
Lot Number10237067 Certificate TypeCertificate of Analysis Date12/04/2022 Product CodeH27201.06,H27201.18
Lot Number10181992 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH27201.06,H27201.18
compliance-icons
Product Identifier
  • 1-Butyl-3-methylimidazolium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/face, hands and anyexposed skin.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1-Butyl-3-methylimidazolium bromide
RUO – Research Use Only
Product Content Correction
Product Title
1-n-Butyl-3-methylimidazolium bromide, 99% >