Få mere at vide
1-Methyluracil, TRC
Artikelnummer. 30180007
Skift visning
Mængde:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180007 1 g
30179810 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30180007 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3381501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 615-77-0
Molekylær formel C5 H6 N2 O2
Molekylvægt (g/mol) 126.11
IUPAC navn 1-methylpyrimidine-2,4-dione
SMIL CN1C=CC(=O)NC1=O
Tekniske data
Kemisk navn eller materiale 1-Methyluracil
Mængde 1 g
InChI formel InChI=1S/C5H6N2O2/c1-7-3-2-4(8)6-5(7)9/h2-3H,1H3,(H,6,8,9)
Formel vægt 126.0429