Få mere at vide
1-Hydroxycyclopentyl 2-chlorophenyl Ketone, TRC
Artikelnummer. 30134183
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134183 100 mg
30136838 250 mg
30006129 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30134183 Leverandør Toronto Research Chemicals Leverandørnr. TRCH949885100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 90717-17-2
Molekylær formel C12 H13 Cl O2
Molekylvægt (g/mol) 224.68
Synonym (2-Chlorophenyl)(1-hydroxycyclopentyl)methanone, 1-Hydroxycyclopentyl 2-chlorophenyl ketone, o-Chlorophenyl 1-hydroxycyclopentyl ketone
IUPAC navn (2-chlorophenyl)-(1-hydroxycyclopentyl)methanone
SMIL OC1(CCCC1)C(=O)c2ccccc2Cl
Tekniske data
Kemisk navn eller materiale diethoxy-(ethylsulfanylmethylsulfanyl)-sulfanylidene-λ^{5}-phosphane
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C12H13ClO2/c13-10-6-2-1-5-9(10)11(14)12(15)7-3-4-8-12/h1-2,5-6,15H,3-4,7-8H2
Formel vægt 224.0604
Procent renhed >95%