Få mere at vide
1-Decylamine Hydrochloride, TRC
Artikelnummer. 30368202
Skift visning
Mængde:
100 mg
250 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30368202 100 mg
30363545 250 mg
30351061 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30368202 Leverandør Toronto Research Chemicals Leverandørnr. TRCD226930100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 143-09-9
Molekylær formel C10H23N . HCl
Molekylvægt (g/mol) 193.76
Synonym 1-Decanamine Hydrochloride, Decanamine Hydrochloride, Decylammonium Chloride, n-Decylamine Hydrochloride, n-Decylammonium Chloride
IUPAC navn decan-1-amine;hydrochloride
SMIL Cl.CCCCCCCCCCN
Tekniske data
Kemisk navn eller materiale 1-Decylamine Hydrochloride
Mængde 100 mg
InChI formel InChI=1S/C10H23N.ClH/c1-2-3-4-5-6-7-8-9-10-11;/h2-11H2,1H3;1H
Formel vægt 193.1597