CAS RN 143-09-9
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
1-Decylamine Hydrochloride, TRC
CAS: 143-09-9 Molekylær formel: C10H23N . HCl Molekylvægt (g/mol): 193.76 Synonym: 1-Decanamine Hydrochloride,Decanamine Hydrochloride,Decylammonium Chloride,n-Decylamine Hydrochloride,n-Decylammonium Chloride IUPAC navn: decan-1-amine;hydrochloride SMIL: Cl.CCCCCCCCCCN