Få mere at vide
1-Chloro-2-(dichlorophenylmethyl)benzene, TRC
Artikelnummer. 30138613
Skift visning
Mængde:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138613 2.5 g
30003470 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30138613 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3656852.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3509-85-1
Molekylær formel C13 H9 Cl3
Molekylvægt (g/mol) 271.57
Synonym (2-Chlorophenyl)phenyldichloromethane
IUPAC navn 1-chloro-2-[dichloro(phenyl)methyl]benzene
SMIL Clc1ccccc1C(Cl)(Cl)c2ccccc2
Tekniske data
Kemisk navn eller materiale (2-Chlorophenyl)phenyldichloromethane
Mængde 2.5 g
InChI formel InChI=1S/C13H9Cl3/c14-12-9-5-4-8-11(12)13(15,16)10-6-2-1-3-7-10/h1-9H
Formel vægt 269.977