missing translation for 'onlineSavingsMsg'
Få mere at vide

1,5-Pentanediyl Bismethanethiosulfonate, TRC

Artikelnummer. 30409839
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30409839 10 mg
30415043 50 mg
30419505 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30409839 Leverandør Toronto Research Chemicals Leverandørnr. TRCP27325010MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 56-00-8
Molekylær formel C7 H16 O4 S4
Molekylvægt (g/mol) 292.46
IUPAC navn 1,5-bis(methylsulfonylsulfanyl)pentane
SMIL CS(=O)(=O)SCCCCCSS(=O)(=O)C

Tekniske data

Kemisk navn eller materiale 1,5-Pentanediyl Bismethanethiosulfonate
Mængde 10 mg
InChI formel InChI=1S/C7H16O4S4/c1-14(8,9)12-6-4-3-5-7-13-15(2,10)11/h3-7H2,1-2H3
Formel vægt 291.9931
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.