Learn More
1,4-Diisopropylcyclohexane, cis + trans, 98%, Thermo Scientific™
400.18 DKK - 1458.67 DKK
Chemical Identifiers
CAS | 22907-72-8 |
---|---|
Molecular Formula | C12H24 |
Molecular Weight (g/mol) | 168.324 |
MDL Number | MFCD00019366 |
InChI Key | GMUJGUXGVJZHSK-UHFFFAOYSA-N |
Synonym | 1,4-diisopropylcyclohexane, 1,4-diisopropyl cyclohexane, 1,4-di-isopropylcyclohexane, 1,4-di propan-2-yl cyclohexane, cyclohexane,1,4-bis 1-methylethyl-,trans, acmc-1cbxy, 1,4-diisopr opylcyclohexane, 1,4-bis methylethyl cyclohexane, 1alpha,4beta-diisopropylcyclohexane, 1,4-diisopropylcyclohexane,cis+trans |
PubChem CID | 140972 |
IUPAC Name | 1,4-di(propan-2-yl)cyclohexane |
SMILES | CC(C)C1CCC(CC1)C(C)C |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
22907-72-8 | |
168.324 | |
GMUJGUXGVJZHSK-UHFFFAOYSA-N | |
140972 | |
CC(C)C1CCC(CC1)C(C)C |
C12H24 | |
MFCD00019366 | |
1,4-diisopropylcyclohexane, 1,4-diisopropyl cyclohexane, 1,4-di-isopropylcyclohexane, 1,4-di propan-2-yl cyclohexane, cyclohexane,1,4-bis 1-methylethyl-,trans, acmc-1cbxy, 1,4-diisopr opylcyclohexane, 1,4-bis methylethyl cyclohexane, 1alpha,4beta-diisopropylcyclohexane, 1,4-diisopropylcyclohexane,cis+trans | |
1,4-di(propan-2-yl)cyclohexane |
Specifications
22907-72-8 | |
C12H24 | |
UN1993 | |
GMUJGUXGVJZHSK-UHFFFAOYSA-N | |
1,4-di(propan-2-yl)cyclohexane | |
140972 | |
98% |
214°C to 216°C | |
MFCD00019366 | |
1,4-diisopropylcyclohexane, 1,4-diisopropyl cyclohexane, 1,4-di-isopropylcyclohexane, 1,4-di propan-2-yl cyclohexane, cyclohexane,1,4-bis 1-methylethyl-,trans, acmc-1cbxy, 1,4-diisopr opylcyclohexane, 1,4-bis methylethyl cyclohexane, 1alpha,4beta-diisopropylcyclohexane, 1,4-diisopropylcyclohexane,cis+trans | |
CC(C)C1CCC(CC1)C(C)C | |
168.324 | |
168.32 | |
1,4-Diisopropylcyclohexane, cis + trans |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures