Få mere at vide
1-(4-Chlorophenyl)-1-phenylethanol, TRC
Artikelnummer. 30133396
Skift visning
Mængde:
25 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133396 1 g
17954695 25 mg
17964265 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133396 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3804751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 59767-24-7
Molekylær formel C14 H13 Cl O
Molekylvægt (g/mol) 232.71
IUPAC navn 1-(4-chlorophenyl)-1-phenylethanol
SMIL CC(O)(c1ccccc1)c2ccc(Cl)cc2
Tekniske data
Kemisk navn eller materiale 1-(4-Chlorophenyl)-1-Phenylethanol
Mængde 1 g
Synonym 1-(4-Chlorophenyl)-1-phenylethanol, : Benzhydrol, 4-chloro-α-methyl- (6CI), (p-Chlorophenyl)methylphenylcarbinol, 1-(4-Chlorophenyl)-1-phenylethanol, 1-(p-Chlorophenyl)-1-phenylethanol, 4-Chloro(α-methyl-α-phenyl)benzenemethanol, EGIS 5927, p-Chloro-α-methylbenzhydrol
InChI formel InChI=1S/C14H13ClO/c1-14(16,11-5-3-2-4-6-11)12-7-9-13(15)10-8-12/h2-10,16H,1H3
Formel vægt 232.0655