Få mere at vide
1-(4-Carboxybutyl)-3,7-dimethylxanthine, TRC
Artikelnummer. 30367004
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367004 10 mg
30352928 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30367004 Leverandør Toronto Research Chemicals Leverandørnr. TRCC17768010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 38975-44-9
Molekylær formel C12H16N4O4
Molekylvægt (g/mol) 280.28
Synonym 1-(4'-Carboxybutyl)-3,7-dimethylxanthine, 2,3,6,7-Tetrahydro-3,7-dimethyl-2,6-dioxo-1H-purine-1-pentanoic Acid, Pentoxifylline M4
IUPAC navn 5-(3,7-dimethyl-2,6-dioxopurin-1-yl)pentanoic acid
SMIL CN1C(=O)N(CCCCC(=O)O)C(=O)c2c1ncn2C
Tekniske data
Kemisk navn eller materiale 1-(4-Carboxybutyl)-3,7-dimethylxanthine
Mængde 10 mg
InChI formel InChI=1S/C12H16N4O4/c1-14-7-13-10-9(14)11(19)16(12(20)15(10)2)6-4-3-5-8(17)18/h7H,3-6H2,1-2H3,(H,17,18)
Formel vægt 280.1172