Få mere at vide
1,4,7-Trioxacyclotridecane-8,13-dione, TRC
Artikelnummer. 17973685
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17973685 100 mg
17971735 1 g
17963435 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17973685 Leverandør Toronto Research Chemicals Leverandørnr. TRCT805500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6607-34-7
Molekylær formel C10 H16 O5
Molekylvægt (g/mol) 216.23
Synonym Adipic Acid Cyclic Diester with Diethylene Glycol, Diethylene Glycol Cyclic Adipate, Cyclic Diethylene Glycol Adipate;
IUPAC navn 1,4,7-trioxacyclotridecane-8,13-dione
SMIL O=C1CCCCC(=O)OCCOCCO1
Tekniske data
Kemisk navn eller materiale 1,4,7-Trioxacyclotridecane-8,13-dione
Renhedsgrad noter GC
Mængde 100 mg
InChI formel InChI=1S/C10H16O5/c11-9-3-1-2-4-10(12)15-8-6-13-5-7-14-9/h1-8H2
Formel vægt 216.0998
Procent renhed >95%