Få mere at vide
1,3-Dibenzylurea, TRC
Artikelnummer. 30132748
Skift visning
Mængde:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132748 1 g
30005395 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30132748 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4185001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1466-67-7
Molekylær formel C15 H16 N2 O
Molekylvægt (g/mol) 240.3
Synonym Lacosamide Impurity C, N,N'-Bis(phenylmethyl)urea, N,N'-Dibenzylurea, sym-Dibenzylurea
IUPAC navn 1,3-dibenzylurea
SMIL O=C(NCc1ccccc1)NCc2ccccc2
Tekniske data
Kemisk navn eller materiale 1,3-Dibenzylurea
Mængde 1 g
InChI formel InChI=1S/C15H16N2O/c18-15(16-11-13-7-3-1-4-8-13)17-12-14-9-5-2-6-10-14/h1-10H,11-12H2,(H2,16,17,18)
Formel vægt 240.1263