Få mere at vide
1,3,5-Tris(4-aminophenyl)benzene, TRC
Artikelnummer. 30240839
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240839 50 mg
30240547 500 mg
30240694 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30240839 Leverandør Toronto Research Chemicals Leverandørnr. TRCT87500850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 118727-34-7
Molekylær formel C24H21N3
Molekylvægt (g/mol) 351.44
IUPAC navn 4-[3,5-bis(4-aminophenyl)phenyl]aniline
SMIL Nc1ccc(cc1)c2cc(cc(c2)c3ccc(N)cc3)c4ccc(N)cc4
Tekniske data
Kemisk navn eller materiale 1,3,5-Tris(4-aminophenyl)benzene
Mængde 50 mg
InChI formel InChI=1S/C24H21N3/c25-22-7-1-16(2-8-22)19-13-20(17-3-9-23(26)10-4-17)15-21(14-19)18-5-11-24(27)12-6-18/h1-15H,25-27H2
Formel vægt 351.1735