Få mere at vide
1,3,5-O-Methylidyne-myo-inositol, TRC
Artikelnummer. 30473237
Skift visning
Mængde:
2.5 g
25 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30473237 2.5 g 2.5g
30477060 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30473237 Leverandør Toronto Research Chemicals Leverandørnr. TRCM3129402.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 98510-20-4
Molekylær formel C7H10O6
Molekylvægt (g/mol) 190.15
Synonym 2,4,10-Trioxatricyclo[3.3.1.13,7]decane, myo-Inositol deriv., myo-Inositol-1,3,5-orthoformate, myo-Inositol Monoorthoformate
SMIL O[C@@H]1[C@H]2O[C@@H]3OC([C@@H](O)C1O3)[C@@H]2O
Tekniske data
Kemisk navn eller materiale 1,3,5-O-Methylidyne-myo-inositol
Mængde 2.5 g
InChI formel InChI=1S/C7H10O6/c8-1-4-2(9)6-3(10)5(1)12-7(11-4)13-6/h1-10H/t1-,2-,3-,4+,5?,6?,7-/m0/s1
Formel vægt 190.0477