Få mere at vide
1-[[(2R,3S)-2-(2,4-Difluorophenyl)-3-methyloxiranyl]methyl]-1H-1,2,4-triazole, TRC
Artikelnummer. 30376503
Skift visning
Mængde:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30376503 500 mg
30370624 5 g
30371107 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30376503 Leverandør Toronto Research Chemicals Leverandørnr. TRCD447815500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 127000-90-2
Molekylær formel C12H11F2N3O
Molekylvægt (g/mol) 251.23
Synonym (2R,3S)-2-(2,4-Difluorophenyl)-3-methyl-[(1H-1,2,4-triazol-1-yl)methyl]oxirane, (2R,3S)-2-(2,4-Difluorophenyl)-3-methyl-2-(1H-1,2,4-triazol-1-yl)methyloxirane
SMIL C[C@@H]1O[C@@]1(Cn2cncn2)c3ccc(F)cc3F
Tekniske data
Kemisk navn eller materiale 1-(((2R,3S)-2-(2,4-Difluorophenyl)-3-methyloxiranyl)methyl)-1H-1,2,4-triazole
Mængde 500 mg
InChI formel InChI=1S/C12H11F2N3O/c1-8-12(18-8,5-17-7-15-6-16-17)10-3-2-9(13)4-11(10)14/h2-4,6-8H,5H2,1H3/t8-,12+/m0/s1
Formel vægt 251.087