Få mere at vide
1,2-Dimethyl-1H-imidazole-5-sulphonyl Chloride, TRC
Artikelnummer. 30004796
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30004796 50 mg
30004716 10 mg
30007852 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30004796 Leverandør Toronto Research Chemicals Leverandørnr. TRCD48758050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 849351-92-4
Molekylær formel C5H7N2O2SCl
Molekylvægt (g/mol) 194.64
SMIL CC1=NC=C(N1C)S(=O)(=O)Cl
Tekniske data
Kemisk navn eller materiale 1,2-Dimethyl-1H-imidazole-5-sulphonyl Chloride
Mængde 50 mg
InChI formel InChI=1S/C5H7ClN2O2S/c1-4-7-3-5(8(4)2)11(6,9)10/h3H,1-2H3
Formel vægt 193.9917