Få mere at vide
1-(2,4-Difluorophenyl)piperazine, 99%, Thermo Scientific™
Artikelnummer. 11358676
Skift visning
Mængde:
1 g
5 g
25 g
Pakningsstørrelse:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11358676 1 g 1g
11368676 5 g 5g
11378676 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 11358676 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. L19393.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 115761-79-0
Molekylær formel C10H12F2N2
Molekylvægt (g/mol) 198.217
MDL nummer MFCD00082588
InChI nøgle CMCSPBOWEYUGHB-UHFFFAOYSA-N
PubChem CID 2734637
IUPAC navn 1-(2,4-difluorophenyl)piperazine
SMIL C1CN(CCN1)C2=C(C=C(C=C2)F)F
Tekniske data
Kemisk navn eller materiale 1-(2,4-Difluorophenyl)piperazine
Smeltepunkt 74°C to 76°C
Kogepunkt 108°C to 111°C (0.2 mmHg)
Mængde 1 g
FN nummer UN2923
Beilstein 8620369
Følsomhed Air Sensitive
Synonym 1-2,4-difluorophenyl piperazine, 1-2,4-difluorophenyl-piperazine, piperazine, 1-2,4-difluorophenyl, 2,4-difluorophenyl piperazine, pubchem9352, acmc-1ca8h, 2,4-difluorophenylpiperazine, 4-2,4-difluorophenyl piperazine, 1-2,4-difluoro-phenyl-piperazine, piperazine,1-2,4-difluorophenyl
Formel vægt 198.22
Procent renhed 99%