Få mere at vide
1-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanone, TRC
Artikelnummer. 30138479
Skift visning
Mængde:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138479 250 mg
30135900 100 mg
17961825 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138479 Leverandør Toronto Research Chemicals Leverandørnr. TRCD440540250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 58905-16-1
Molekylær formel C10 H7 Cl2 N3 O
Molekylvægt (g/mol) 256.088
Synonym 1-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanone, 1-(2,4-Dichloro-phenyl)-2-[1,2,4]triazol-1-yl-ethanone, 1-(2,4-Dichlorophenyl)-2-(1,2,4-triazol-1-yl)ethanone, 1-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-yl)-1-ethanone, 2',4'-Dichloro-2-(1,2,4-triazol-1-yl)acetophenone, 2',4'-Dichloro-2-(1H-1,2,4-triazol-1-yl)acetophenone, 1-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanone, 1-(2,4-Dichloro-phenyl)-2-[1,2,4]triazol-1-yl-ethanone, 1-(2,4-Dichlorophenyl)-2-(1,2,4-triazol-1-yl)ethanone, 1-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-yl)-1-ethanone, 2',4'-Dichloro-2-(1,2,4-t
IUPAC navn 1-(2,4-dichlorophenyl)-2-(1,2,4-triazol-1-yl)ethanone
SMIL Clc1ccc(C(=O)Cn2cncn2)c(Cl)c1
Tekniske data
Kemisk navn eller materiale 1-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanone
Mængde 250 mg
InChI formel InChI=1S/C10H7Cl2N3O/c11-7-1-2-8(9(12)3-7)10(16)4-15-6-13-5-14-15/h1-3,5-6H,4H2
Formel vægt 254.997