Få mere at vide
1,2,3,4,5-Pentabromo-6-(2,3,5,6-tetrabromophenoxy)benzene, TRC
Artikelnummer. 30134214
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134214 100 mg
30010326 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134214 Leverandør Toronto Research Chemicals Leverandørnr. TRCP237850100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 437701-78-5
Molekylær formel C12 H Br9 O
Molekylvægt (g/mol) 880.27
Synonym Benzene, 1,2,3,4,5-pentabromo-6-(2,3,5,6-tetrabromophenoxy)-, Benzene, pentabromo(2,3,5,6-tetrabromophenoxy)- (9CI), 1,2,3,4,5-Pentabromo-6-(2,3,5,6-tetrabromophenoxy)benzene, 2,2',3,3',4,5,5',6,6'-Nonobromodiphenyl ether, BDE 208, PBDE 208
IUPAC navn 1,2,3,4,5-pentabromo-6-(2,3,5,6-tetrabromophenoxy)benzene
SMIL Brc1cc(Br)c(Br)c(Oc2c(Br)c(Br)c(Br)c(Br)c2Br)c1Br
Tekniske data
Kemisk navn eller materiale PBDE 208
Mængde 100 mg
InChI formel InChI=1S/C12HBr9O/c13-2-1-3(14)5(16)11(4(2)15)22-12-9(20)7(18)6(17)8(19)10(12)21/h1H
Formel vægt 871.2678