Få mere at vide
1-[2-(2,4-Dichlorophenyl)-2-[(3,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole, TRC
Artikelnummer. 30005515
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30005515 10 mg
30011881 100 mg
30010462 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30005515 Leverandør Toronto Research Chemicals Leverandørnr. TRCD43203810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 47447-55-2
Molekylær formel C18H14Cl4N2O
Molekylvægt (g/mol) 416.13
Synonym Miconazole EP Impurity F;
SMIL Clc1ccc(C(Cn2ccnc2)OCc3ccc(Cl)c(Cl)c3)c(Cl)c1
Tekniske data
Kemisk navn eller materiale 1-[2-(2,4-Dichlorophenyl)-2-[(3,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C18H14Cl4N2O/c19-13-2-3-14(16(21)8-13)18(9-24-6-5-23-11-24)25-10-12-1-4-15(20)17(22)7-12/h1-8,11,18H,9-10H2
Formel vægt 413.986
Procent renhed >95%