Få mere at vide
1-(1-Phenylpropan-2-yl)-1H-pyrrole-2-carboxylic Acid (>90%), TRC
Artikelnummer. 30009344
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009344 10 mg
30008593 50 mg
30004227 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009344 Leverandør Toronto Research Chemicals Leverandørnr. TRCB52255810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1432680-68-6
Molekylær formel C14 H15 N O2
Molekylvægt (g/mol) 229.27
Synonym 1-(1-Methyl-2-phenylethyl)-1H-pyrrole-2-carboxylic Acid
IUPAC navn 1-(1-methyl-2-phenyl-ethyl)pyrrole-2-carboxylic acid
SMIL CC(Cc1ccccc1)n2cccc2C(=O)O
Tekniske data
Kemisk navn eller materiale 1-(1-Phenylpropan-2-yl)-1H-pyrrole-2-carboxylic acid
Mængde 10 mg
InChI formel InChI=1S/C14H15NO2/c1-11(10-12-6-3-2-4-7-12)15-9-5-8-13(15)14(16)17/h2-9,11H,10H2,1H3,(H,16,17)
Formel vægt 229.1103