missing translation for 'onlineSavingsMsg'
Få mere at vide

1,1,1,3,3,3-Hexafluoro-2-(fluoromethoxy)propane, TRC

Artikelnummer. 30136079
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136079 5 g
30008797 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.
Artikelnummer. 30136079 Leverandør Toronto Research Chemicals Leverandørnr. TRCH2910505G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product contains a substance listed in Annex I to EU Regulation No. 517/2014 on fluorinated greenhouse gases, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 28523-86-6
Molekylær formel C4 H3 F7 O
Molekylvægt (g/mol) 200.05
Synonym Sevoflurane, 1,1,1,3,3,3-Hexafluoro-2-(fluoromethoxy)propane, Ether, fluoromethyl 2,2,2-trifluoro-1-(trifluoromethyl)ethyl (8CI), 347mmzEβγ, Fluoromethyl 1,1,1,3,3,3-hexafluoro-2-propyl ether, Fluoromethyl 1,1,1,3,3,3-hexafluoroisopropyl ether, Fluoromethyl 2,2,2-trifluoro-1-(trifluoromethyl)ethyl ether, MR 6S4, R-E 347, Sevocris, Sevoflo, Sevofluran, Sevoflurane, Sevofrane, Sevorane, Ultane
IUPAC navn 1,1,1,3,3,3-hexafluoro-2-(fluoromethoxy)propane
SMIL FCOC(C(F)(F)F)C(F)(F)F

Tekniske data

Kemisk navn eller materiale Sevoflurane
Mængde 5 g
InChI formel InChI=1S/C4H3F7O/c5-1-12-2(3(6,7)8)4(9,10)11/h2H,1H2
Formel vægt 200.0072
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.