CAS RN 94-97-3
CAS RN 94-97-3
IUPAC Navn:
5-chloro-2H-1,2,3-benzotriazole
Synonymer:
5-chloro-2H-1,2,3-benzotriazole
Molekylvægt (g/mol):
153.57
Molekylformel:
C6H4ClN3
InChi Key:
PZBQVZFITSVHAW-UHFFFAOYSA-N
SMILES:
ClC1=CC2=NNN=C2C=C1
1
–
3
af
3
Resultater
1
5-Chlorobenzotriazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-Chlorobenzotriazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5-Chlorobenzotriazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.