accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 91983-14-1

HOBOHBr

CAS RN 91983-14-1

IUPAC Name: [2-(bromomethyl)phenyl]boronic acid
Synonyms: 2-(bromomethyl)phenylboronic acid
Molecular Weight (g/mol): 214.85
Molecular Formula: C7H8BBrO2
InChi Key: MYVJCOQGXCONPE-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC=C1CBr
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.