accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 918523-99-6

FHOFO

CAS RN 918523-99-6

IUPAC Name: 2,5-difluoro-4-hydroxybenzaldehyde
Synonyms: 2,5-difluoro-4-hydroxybenzaldehyde
Molecular Weight (g/mol): 158.1
Molecular Formula: C7H4F2O2
InChi Key: FFPHRYLGIPAZBL-UHFFFAOYSA-N
SMILES: OC1=C(F)C=C(C=O)C(F)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.