accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 903576-44-3

NOClHCl

CAS RN 903576-44-3

IUPAC Name: 1-{3-[3-(4-chlorophenyl)propoxy]propyl}piperidine hydrochloride
Synonyms: pitolisant
Molecular Weight (g/mol): 332.31
Molecular Formula: C17H27Cl2NO
InChi Key: XLFKECRRMPOAQS-UHFFFAOYSA-N
SMILES: Cl.ClC1=CC=C(CCCOCCCN2CCCCC2)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.