CAS RN 89705-21-5
CAS RN 89705-21-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express APNEA, MedChemExpress
MedChemExpress APNEA (N6-[2-(4-Aminophenyl)ethyl]adenosine) is a potent, non-selective A3 adenosine receptor agonist.
