CAS RN 88634-80-4
CAS RN 88634-80-4
IUPAC Name:
2-ethyl-5-methyl-1H-imidazole-4-carbaldehyde
Synonyms:
2-ethyl-5-methyl-1H-imidazole-4-carbaldehyde
Molecular Weight (g/mol):
138.17
Molecular Formula:
C7H10N2O
InChi Key:
RLBMKAXUSJOVJI-UHFFFAOYSA-N
SMILES:
CCC1=NC(C=O)=C(C)N1
1
–
1
of
1
results
1
Thermo Scientific Maybridge 2-Ethyl-4-methyl-1H-imidazole-5-carbaldehyde, 97%, Thermo Scientific™
CAS: 88634-80-4 Molecular Formula: C7H10N2O Molecular Weight (g/mol): 138.17 MDL Number: MFCD00173727 InChI Key: RLBMKAXUSJOVJI-UHFFFAOYSA-N PubChem CID: 2795884 IUPAC Name: 2-ethyl-5-methyl-1H-imidazole-4-carbaldehyde SMILES: CCC1=NC(C=O)=C(C)N1