CAS RN 874-23-7
CAS RN 874-23-7
IUPAC Name:
2-acetylcyclohexan-1-one
Synonyms:
2-acetylcyclohexan-1-one
Molecular Weight (g/mol):
140.18
Molecular Formula:
C8H12O2
InChi Key:
OEKATORRSPXJHE-UHFFFAOYNA-N
SMILES:
CC(=O)C1CCCCC1=O
1
–
1
of
1
results
1
Thermo Scientific Acros 2-Acetylcyclohexanone, 97%
CAS: 874-23-7 Molecular Formula: C8H12O2 Molecular Weight (g/mol): 140.18 MDL Number: MFCD00001633 InChI Key: OEKATORRSPXJHE-UHFFFAOYNA-N Synonym: 2-acetylcyclohexanone,cyclohexanone, 2-acetyl,2-acetyl-1-cyclohexanone,.alpha.-acetylcyclohexanone,2-acetyl cyclohexanone,2-acetyl-cyclohexanone,cyclohexanone,2-acetyl,1-acetyl-2-oxocyclohexane,alpha-acetylcyclohexanone,acetylcyclohexanone PubChem CID: 13400 IUPAC Name: 2-acetylcyclohexan-1-one SMILES: CC(=O)C1CCCCC1=O