CAS RN 869748-10-7
CAS RN 869748-10-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express A-770041, MedChemExpress
MedChemExpress A-770041 is selective and orally active Src-family Lck inhibitor; A-770041 is a 147 nM inhibitor of Lck (1 mM ATP) and is 300-fold selective against Fyn, the other Src family kinase involved in T-cell signaling.
