CAS RN 79622-59-6
CAS RN 79622-59-6
IUPAC Name:
3-chloro-N-[3-chloro-2,6-dinitro-4-(trifluoromethyl)phenyl]-5-(trifluoromethyl)pyridin-2-amine
Synonyms:
fluazinam
Molecular Weight (g/mol):
465.09
Molecular Formula:
C13H4Cl2F6N4O4
InChi Key:
UZCGKGPEKUCDTF-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC(=C(Cl)C(=C1NC1=NC=C(C=C1Cl)C(F)(F)F)[N+]([O-])=O)C(F)(F)F
1
–
2
of
2
results
2
Medchem Express Fluazinam, MedChemExpress
MedChemExpress Fluazinam is a broad spectrum pyridinamine fungal inhibitor.
