accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 65-29-2

IIICH3NCH3H3COONH3CH3CH3CONCH3CH3H3C

CAS RN 65-29-2

IUPAC Name: (2-{2,3-bis[2-(triethylazaniumyl)ethoxy]phenoxy}ethyl)triethylazanium triiodide
Synonyms: gallamine triethiodide (inn)
Molecular Weight (g/mol): 891.54
Molecular Formula: C30H60I3N3O3
InChi Key: REEUVFCVXKWOFE-UHFFFAOYSA-K
SMILES: [I-].[I-].[I-].CC[N+](CC)(CC)CCOC1=CC=CC(OCC[N+](CC)(CC)CC)=C1OCC[N+](CC)(CC)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.