CAS RN 6133-30-8
CAS RN 6133-30-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Guanidinosuccinic acid, MedChemExpress
MedChemExpress Guanidinosuccinic acid is a nitrogenous metabolite.
