accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 563-46-2

H3CCH3H2C

CAS RN 563-46-2

IUPAC Name: 2-methylbut-1-ene
Synonyms: methylbutenes
Molecular Weight (g/mol): 70.14
Molecular Formula: C5H10
InChi Key: MHNNAWXXUZQSNM-UHFFFAOYSA-N
SMILES: CCC(C)=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.