accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 49608-01-7

H3COOClN

CAS RN 49608-01-7

IUPAC Name: ethyl 6-chloropyridine-3-carboxylate
Synonyms: ethyl 6-chloropyridine-3-carboxylate
Molecular Weight (g/mol): 185.61
Molecular Formula: C8H8ClNO2
InChi Key: ILDJJTQWIZLGPO-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(Cl)N=C1
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.