CAS RN 4233-96-9
CAS RN 4233-96-9
IUPAC Name:
(2S,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-3-yl 3,4,5-trihydroxybenzoate
Synonyms:
gallocatechin gallate
Molecular Weight (g/mol):
458.38
Molecular Formula:
C22H18O11
InChi Key:
WMBWREPUVVBILR-NQIIRXRSSA-N
SMILES:
OC1=CC(O)=C2C[C@@H](OC(=O)C3=CC(O)=C(O)C(O)=C3)[C@@H](OC2=C1)C1=CC(O)=C(O)C(O)=C1
1
–
2
of
2
results
2
Medchem Express (-)-Gallocatechin gallate, MedChemExpress
MedChemExpress (-)-Gallocatechin gallate is the polyphenol isolated from tea, with cancer-preventive activities.
